EMAIL THIS PAGE TO A FRIEND
Journal of cosmetic science
Synthesis of new fragrances from 2-methylfuran. Part III.
PMID 14605690
Abstract
The synthesis of new 2-methylfuran-based alcohols is described. The odor and physicochemical characteristics of all new compounds are also presented.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M46845
2-Methylfuran, contains 200-400 BHT as stabilizer, 99%
C5H6O