EMAIL THIS PAGE TO A FRIEND
Journal of microbiological methods
Use of 3-hydroxyphenylacetylene for activity-dependent, fluorescent labeling of bacteria that degrade toluene via 3-methylcatechol.
PMID 14607424
Abstract
3-hydroxyphenylacetylene (3-HPA) served as a novel, activity-dependent, fluorogenic and chromogenic probe for bacterial enzymes known to degrade toluene via meta ring fission of the intermediate, 3-methylcatechol. By this direct physiological analysis, cells grown with an aromatic substrate to induce the synthesis of toluene-degrading enzymes were fluorescently labeled.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
632023
3-Hydroxyphenylacetylene, ≥95.0%
C8H6O
M34006
3-Methylcatechol, 98%
C7H8O2