EMAIL THIS PAGE TO A FRIEND
Die Naturwissenschaften
Methyl 2-(methylthio)benzoate: the unique sulfur-containing sex pheromone of Phyllophaga crinita.
PMID 14610650
Abstract
The female-produced sex pheromone of Phyllophaga crinita (Burmeister) (Coleoptera: Scarabaeidae: Melolonthinae; the adult has no common name) is identified as methyl 2-(methylthio)benzoate. This is the first identification of a sulfur-containing, long-distance, female-produced sex attractant from any insect taxa. The root-feeding larvae of this species are serious pests in many crops in Texas and Mexico. In field tests, many P. crinita males were captured in traps baited with the authentic compound. Interestingly, a heteroatom analog, methyl 2-methoxybenzoate, also captured P. crinita males, but only at a dose 10,000 times higher than the lowest tested dose of the authentic pheromone.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
451436
3-Mercaptobenzoic acid, 95%
C7H6O2S
JRD0440
Methyl 4-(methylthio)benzoate, AldrichCPR
C9H10O2S