EMAIL THIS PAGE TO A FRIEND
European psychiatry : the journal of the Association of European Psychiatrists
Lithium augmentation in venlafaxine non-responders: an open study.
PMID 14611927
Abstract
Thirteen major depressive patients not responding to a 4-week venlafaxine 300 mg treatment were eligible for a 4-week open trial of lithium addition. Two patients had to stop lithium for a possible moderate serotonin syndrome and five patients became responders, including one dramatic response and two semi-rapid responses.