EMAIL THIS PAGE TO A FRIEND
The Journal of clinical investigation
Pseudomonas aeruginosa quorum sensing as a potential antimicrobial target.
PMID 14617745
Abstract
Pseudomonas aeruginosa has two complete quorum-sensing systems. Both of these systems have been shown to be important for Pseudomonas virulence in multiple models of infection. Thus, these systems provide unique targets for novel antimicrobial drugs.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
68749
3-Oxo-N-(2-oxocyclohexyl)dodecanamide, ≥95.0%
C18H31NO3