EMAIL THIS PAGE TO A FRIEND
Cardiovascular journal of South Africa : official journal for Southern Africa Cardiac Society [and] South African Society of Cardiac Practitioners
COMET challenges paradigm of ACE inhibition in mild heart failure.
PMID 14619868