EMAIL THIS PAGE TO A FRIEND
Federal Register
International Conference on Harmonisation; final recommendations on the revision of the permitted daily exposures for two solvents, n-methylpyrrolidone and tetrahydrofuran, according to the maintenance procedures for the guidance Q3C Impurities: Residual Solvents; Availability. Notice.
PMID 14619948