EMAIL THIS PAGE TO A FRIEND
Experimental and toxicologic pathology : official journal of the Gesellschaft fur Toxikologische Pathologie
Influence of age on iminodipropionitrile-induced vestibular and neurobehavioral toxicities in rats.
PMID 14620540
Abstract
A direct association between aging and drug-induced dyskinesia has been reported by several investigators. Iminiodipropionitrile (IDPN), a prototype nitrile compound produces a motor syndrome in rodents, which resembles neuroleptic drug induced dyskinesia. In this investigation attempt has been made to study the effect of age on IDPN induced vestibular hair cell degeneration and resulting dyskinetic syndrome. Male Wistar rats aged 3, 6 and 12 weeks received IDPN in the doses of 0, 200 and 400 mg/kg, intraperitoneally for 3 consecutive days. IDPN-induced dyskinesia was assessed using a behavioral testing battery on days 3, 4, 5, 6, 7, 14, 21 and 28. The rats were sacrificed on day 28; temporal bones were excised for vestibular histopathology and sera were collected for measuring the indices of oxidative stress (glutathione and conjugated dienes). IDPN in the dose of 200 mg/kg produced dyskinesia in 12 weeks old rats, but failed to do so in 3 and 6 weeks old rats. The high dose of IDPN (400 mg/kg) caused dyskinesia in all age groups, however, its onset and severity were age-dependent. Older rats showed an early onset and significantly high incidence of dyskinesia as compared to younger rats. The susceptibility of rats to IDPN-induced behavioral deficits was proportional to oxidative stress and degeneration of sensory hair cells in the crista ampullaris.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
317306
3,3′-Iminodipropionitrile, technical grade, 90%
C6H9N3