EMAIL THIS PAGE TO A FRIEND
The European Journal of Neuroscience
Neurons of the rat preoptic area and the raphe pallidus nucleus innervating the brown adipose tissue express the prostaglandin E receptor subtype EP3.
PMID 14622218