EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
New antibacterial tetrahydro-4(2H)-thiopyran and thiomorpholine S-oxide and S,S-dioxide phenyloxazolidinones.
PMID 14623003
Abstract
Combinatorial libraries of N-acylated 5-(S)-aminomethyloxazolidinone derivatives of S-oxide and S,S-dioxide tetrahydro-4(2H)-thiopyranyl and thiomorpholine phenyloxazolidinone series have been synthesized on a solid phase and evaluated for antimicrobial activity. Several novel potent leads have been identified, including orally active oxazolidinones with enhanced activity against respiratory tract infection pathogens Haemophilus influenzae and Moraxella catarrhalis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
196274
Thiomorpholine, 98%
C4H9NS
88345
Thiomorpholine hydrochloride, ≥98.0% (AT)
C4H9NS · HCl