EMAIL THIS PAGE TO A FRIEND
Toxicology
Acute renal and hepatic toxicity of 2-haloanilines in Fischer 344 rats.
PMID 1462350
Abstract
Aniline and its halogenated derivatives are widely used as chemical intermediates. The purpose of this study was to determine the hepatotoxic and nephrotoxic potential of the 2-haloanilines. Male Fischer 344 rats (n > or = 4) were injected (i.p.) with 1.0 or 1.25 mmol/kg of: aniline (A), 2-fluoroaniline (2-FA), 2-chloroaniline (2-ClA), 2-bromoaniline (2-BrA), 2-iodoaniline (2-IA) or vehicle (0.9% saline, 2.5 ml/kg). All compounds were injected as hydrochloride salts. Renal and hepatic function was monitored 24 h after treatment. All of the 2-haloanilines induced oliguria, diminished kidney weight, tubular casts and decreased renal cortical slice accumulation of organic anions. Blood urea nitrogen (BUN) levels were increased (P < 0.05) by treatment with 1.0 or 1.25 mmol/kg of 2-FA, 2-ClA or 2-BrA. Hepatic alterations were also observed and characterized by elevated plasma ALT/GPT activity and altered morphology in the centrilobular region. The nephrotoxic and hepatotoxic potentials were similar among the 2-haloanilines but aniline was less toxic than its 2-halo derivatives. These results demonstrated that halogen substitution at the 2-position of aniline increased hepatic and renal toxicity. However, the severity of toxicity was not influenced by the nature of the halogen substituent.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F3401
2-Fluoroaniline, ≥99%
C6H6FN