EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences
Microdialysis for the determination of acetaldehyde and ethanol concentrations in the striatum of freely moving rats.
PMID 14630370
Abstract
The subject of the present paper is the simultaneous determination of ethanol (EtOH) and acetaldehyde (AcH) concentrations in the striatum of freely moving rats using an in vivo microdialysis followed by head-space gas chromatography (GC). Major operation conditions of GC were as follows: column, injector and detector temperatures 90, 110 and 200 degrees C, respectively; Supelcowax wide bore capillary column (60 m length, 0.53 mm i.d., 2 microm film thickness); carrier gas, nitrogen; flow rate, 20 ml/min. The recovery of EtOH and AcH at a concentration 40 mM and 250 microM, respectively, by microdialysis showed a maximum of 83.8+/-12.2 and 51.2+/-6.5%, respectively, at a flow rate of 0.8 microl/min. A good linear calibration curve in the concentration range of 5-50 mM for EtOH (r=0.998), and 10-250 microM for AcH (r=0.988) was observed. Microdialysates were collected for 10 min each after insertion of probe into the striatum. Rats were treated with cyanamide (100mg/kg, a potent aldehyde dehydrogenase inhibitor) and 60 min later with EtOH (1g/kg) intraperitoneally. A 10 min sample was about 8 microl. This volume was mixed with 40 microl of 0.002% t-butanol as an internal standard in 0.6N perchloric acid, and then analyzed by head-space GC method. The peak EtOH and AcH concentrations in the striatal dialysates reached maximum at 30 min, and then gradually decreased. This method represents a reasonable tool to quantify in vivo both AcH and EtOH levels simultaneously in rat brain.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
25026-U
SUPELCOWAX® 10 Capillary GC Column, L × I.D. 10 m × 0.10 mm, df 0.10 μm
24343
SUPELCOWAX® 10 Capillary GC Column, L × I.D. 15 m × 0.10 mm, df 0.10 μm
24169
SUPELCOWAX® 10 Capillary GC Column, L × I.D. 30 m × 0.20 mm, df 0.20 μm
24077
SUPELCOWAX® 10 Capillary GC Column, L × I.D. 15 m × 0.25 mm, df 0.25 μm
24079
SUPELCOWAX® 10 Capillary GC Column, L × I.D. 30 m × 0.25 mm, df 0.25 μm
24081
SUPELCOWAX® 10 Capillary GC Column, L × I.D. 60 m × 0.25 mm, df 0.25 μm
23308-U
SUPELCOWAX® 10 Capillary GC Column, df 0.25 μm
24284
SUPELCOWAX® 10 Capillary GC Column, L × I.D. 30 m × 0.25 mm, df 0.50 μm
24078
SUPELCOWAX® 10 Capillary GC Column, L × I.D. 15 m × 0.32 mm, df 0.25 μm
24080-U
SUPELCOWAX® 10 Capillary GC Column, L × I.D. 30 m × 0.32 mm, df 0.25 μm
24082
SUPELCOWAX® 10 Capillary GC Column, L × I.D. 60 m × 0.32 mm, df 0.25 μm
24084
SUPELCOWAX® 10 Capillary GC Column, L × I.D. 30 m × 0.32 mm, df 0.50 μm
24085-U
SUPELCOWAX® 10 Capillary GC Column, L × I.D. 60 m × 0.32 mm, df 0.50 μm
24211
SUPELCOWAX® 10 Capillary GC Column, L × I.D. 30 m × 0.32 mm, df 1.00 μm
24212
SUPELCOWAX® 10 Capillary GC Column, L × I.D. 60 m × 0.32 mm, df 1.00 μm
25325
SUPELCOWAX® 10 Capillary GC Column, L × I.D. 30 m × 0.53 mm, df 0.50 μm
25300-U
SUPELCOWAX® 10 Capillary GC Column, L × I.D. 15 m × 0.53 mm, df 1.00 μm
25301-U
SUPELCOWAX® 10 Capillary GC Column, L × I.D. 30 m × 0.53 mm, df 1.00 μm
25391
SUPELCOWAX® 10 Capillary GC Column, L × I.D. 60 m × 0.53 mm, df 1.00 μm
25375-U
SUPELCOWAX® 10 Capillary GC Column, L × I.D. 30 m × 0.53 mm, df 2.00 μm
25376
SUPELCOWAX® 10 Capillary GC Column, L × I.D. 60 m × 0.53 mm, df 2.00 μm