EMAIL THIS PAGE TO A FRIEND
Journal of magnetic resonance (San Diego, Calif. : 1997)
Compensating for pulse imperfections in REDOR.
PMID 14643704
Abstract
Rotational-echo double resonance (REDOR) is a magic-angle spinning technique for measuring heteronuclear dipolar couplings. Rotor-synchronized pi pulses recouple the dipolar interaction. The accuracy of a REDOR determination of distance or orientation depends totally on the quality of the dephased (recoupled) and full-echo spectra. We present a scheme for measuring and compensating for the effects of pulse imperfections in REDOR experiments. No assumptions are made about the quality of the pi pulses, and no pulses are added or taken away in implementing the compensation for incomplete REDOR dephasing by imperfect pi pulses.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
372455 Graphite, fluorinated, polymer, extent of labeling: >61 wt. % F