EMAIL THIS PAGE TO A FRIEND
The Journal of clinical endocrinology and metabolism
Structure of the human glucokinase gene and identification of a missense mutation in a Japanese patient with early-onset non-insulin-dependent diabetes mellitus.
PMID 1464666
Abstract
Glucokinase is thought to play a glucose-sensor role in the pancreas, and abnormalities in its structure, function, and regulation can induce diabetes. We isolated the human glucokinase gene, and determined its genomic structure including exon-intron boundaries. Structure of the glucokinase gene in human was very similar to that in rat. Then, by screening Japanese diabetic patients using polymerase chain reaction--single strand conformation polymorphism (PCR-SSCP) and direct-sequencing strategies, we identified a missense mutation substituting arginine (AGG) for glycine (GGG) at position 261 in exon 7 of the glucokinase gene in a patient with early-onset non-insulin-dependent diabetes (NIDDM).
Related Materials