EMAIL THIS PAGE TO A FRIEND
Journal of inorganic biochemistry
Peroxidase-catalyzed co-oxidation of 3,3',5,5'-tetramethylbenzidine in the presence of substituted phenols and their polydisulfides.
PMID 14659626
Abstract
The steady-state kinetics of the horseradish peroxidase (HRP)-catalyzed oxidation of 3,3',5,5'-tetramethylbenzidine (TMB) has been studied in the presence of 2-amino-4-nitrophenol (ANP), gallic acid (GA) or 4,4'-dihydroxydiphenylsulfone (DDS) and their polydisulfides poly(ADSNP), poly(DSGA), poly(DSDDS) at 20 degrees C in 10 mM phosphate buffer, pH 6.4, supplemented with 5-10% dimethylformamide. The second-order rate constants for the reactions of ANP, GA, poly(DSGA) and poly(DSDDS) with HRP-Compound I (k2) and Compound II (k3) have been determined at 25 degrees C in 10 mM phosphate buffer, pH 6.0 by stopped-flow spectrophotometry. ANP, GA and their polydisulfides strongly inhibited HRP-catalyzed TMB oxidation. Inhibition constants (Ki) and stoichiometric coefficients of inhibition (f) have been determined for these reactions. The most effective inhibitor was poly(DSGA) (Ki=1.3 microM, f=35.6). The oxidation of substrate pairs by HRP, i.e., TMB-DDS and TMB-poly(DSDDS) at pH 7.2 resulted in a approximately 8- and approximately 12-fold stimulation of TMB oxidation rates, respectively. The mechanisms of the HRP-catalyzed co-oxidation of TMB-phenol pairs are discussed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A70402
2-Amino-4-nitrophenol, 96%
C6H6N2O3