EMAIL THIS PAGE TO A FRIEND
Archiwum medycyny sadowej i kryminologii
[Suicide by propane-butane inhalation: a case report and literature review].
PMID 14669660
Abstract
A case of suicide of a 26-year old woman by propane-butane inhalation is presented with a description of the circumstances, postmortem pathomorphological and toxicological findings. The literature relating to deaths due to inhalation of these gaseous substances was briefly reviewed.
Related Materials