EMAIL THIS PAGE TO A FRIEND
Drug design and discovery
Structure-based design of novel inhibitors of 3-deoxy-D-manno-octulosonate 8-phosphate synthase.
PMID 14675946
Abstract
3-Deoxy-D-manno-octulosonate 8-phosphate (KDO8P) is the phosphorylated precursor of KDO, an essential sugar of the lipopolysaccharide of Gram negative bacteria. KDO8P is produced by a specific synthase (KDO8PS) by condensing arabinose 5-phosphate (A5P) and phosphoenolpyruvate (PEP), with release of inorganic phosphate. As KDO8PS is present in bacteria and plants, but not in mammalian cells, and mutations that inactivate KDO8PS also block cell replication, KDO8PS is a promising target for the design of new antimicrobials that act by blocking lipopolysaccharide biosynthesis. Previous studies have shown that a compound mimicking an intermediate of the condensation reaction is a good ligand and a powerful inhibitor. Here we report on the crystallographic investigation of the binding to KDO8PS of new derivatives of this original inhibitor. The structures of the enzyme in complex with these compounds, and also with the PEP analogs, 2-phosphoglyceric acid (2-PGA) and Z-methyl-PEP, point to future strategies for the design of novel inhibitors of KDO8PS.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
19710
L-2-Phosphoglyceric acid disodium salt hydrate, ≥80% (CE)
C3H5Na2O7P · xH2O