EMAIL THIS PAGE TO A FRIEND
Journal of lipid research
A comparison of the metabolism of eighteen-carbon 13C-unsaturated fatty acids in healthy women.
PMID 14679164
Abstract
Altered use of different dietary fatty acids may contribute to several chronic diseases, including obesity, noninsulin-dependent diabetes mellitus, and cardiovascular disease. However, few comparative data are available to support this link, so the goal of the present study was to compare the metabolism of [(13)C]oleate, [(13)C]alpha-linolenate, [(13)C]elaidate, and [(13)C]linoleate through oxidation and incorporation into plasma lipid fractions and adipose tissue. Each tracer was given as a single oral bolus to six healthy women. Samples were collected over 8 days, and (13)C was analyzed using isotope ratio mass spectrometry. At 9 h postdose, cumulative oxidation was similar for [(13)C]elaidate, [(13)C]oleate, and [(13)C]alpha-linolenate (19 +/- 1%, 20 +/- 4%, and 19 +/- 3% dose, respectively). Significantly lower oxidation of [(13)C]linoleate (12 +/- 4% dose; P < 0.05) was accompanied by its higher incorporation into plasma phospholipids and cholesteryl esters. Abdominal adipose tissue was enriched with [(13)C]alpha-linolenate, [(13)C]elaidate, or [(13)C]linoleate within 6 h. The percentage linoleate in plasma phospholipids correlated positively with [(13)C]linoleate and [(13)C]elaidate oxidation, indicating a potential role of background diet. Conversion of [(13)C]linoleate and [(13)C]alpha-linolenate to longer chain polyunsaturates was a quantitatively minor route of utilization.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E4637
Elaidic acid, ≥99.0% (GC)
C18H34O2
45089
Elaidic acid, analytical standard
C18H34O2