EMAIL THIS PAGE TO A FRIEND
Chemistry (Weinheim an der Bergstrasse, Germany)
Separated and aligned molecular fibres in solid state self-assemblies of cyclodextrin [2]rotaxanes.
PMID 14679509
Abstract
The conformations of two [2]rotaxanes, each comprising alpha-cyclodextrin as the rotor, a stilbene as the axle and 2,4,6-trinitrophenyl substituents as the capping groups, have been examined in solution and in the solid state, using (1)H NMR spectroscopy and X-ray crystallography, respectively. In solution, introducing substituents onto the stilbene prevents the cyclodextrin from being localized over one end of the axle. Instead the cyclodextrin moves back and forth along the substituted stilbene. In the solid state, the axles of the rotaxanes form extended molecular fibres that are separated from each other and aligned along a single axis. The molecular fibres are strikingly similar to those formed by the axle component of one of the rotaxanes in the absence of the cyclodextrin, but in the latter case they are neither separated nor all aligned.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
137790
5-Methyl-2-nitrophenol, 97%
C7H7NO3