EMAIL THIS PAGE TO A FRIEND
Chemistry (Weinheim an der Bergstrasse, Germany)
Installation of a ratchet tooth and pawl to restrict rotation in a cyclodextrin rotaxane.
PMID 14679510
Abstract
Eight new [2]rotaxanes have been prepared, incorporating an alpha-cyclodextrin as the rotor, a stilbene as the axle, and trinitrophenyl substituents as capping groups. Strategies have been devised to elaborate these by linking the rotor to the axle, to produce two new [1]rotaxanes. Rotational motion in a selection of these rotaxanes has been investigated through the application of two-dimensional NMR spectroscopy by performing TOCSY, DQF-COSY, ROESY and HMQC experiments. This has shown that a methoxyl group incorporated on the stilbene and a succinamide joining the stilbene and the cyclodextrin behave analogously to a ratchet tooth and pawl, respectively, to restrict rotation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
124753
Succinamide, 98%
C4H8N2O2