EMAIL THIS PAGE TO A FRIEND
Chronobiology international
Lifetimes of mRNAs for clock-regulated proteins in a dinoflagellate.
PMID 14680137
Abstract
Both pulsed and continuous applications of the RNA polymerase II inhibitor thiolutin cause a dramatic but reversible loss of bioluminescence and its overt rhythmicity in cells of the dinoflagellate Lingulodinium polyedrum (formerly Gonyaulax polyedra). Such cells remain alive, and the rhythm resumes after an interval, the length of which depends on the concentration of thiolutin used. The period and phase of the resumed rhythm were not systematically altered following such treatments, and the effects were not different at different circadian phases. For three different genes, luciferin binding protein (lbp), luciferase (lcf), and glyceraldehyde-3-phosphate dehydrogenase (gapdh), which are circadian-regulated at the level of translation, the amounts of their mRNAs were determined by Northern blots for times up to 12.5 h following the addition of 1.5 microM thiolutin. Consistent with previous reports that their abundances do not change with circadian time, their levels remained high for several hours after thiolutin addition, but then did diminish.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T3450
Thiolutin, from Streptomyces luteosporeus, ≥95% (HPLC)
C8H8N2O2S2