EMAIL THIS PAGE TO A FRIEND
Zhonghua liu xing bing xue za zhi = Zhonghua liuxingbingxue zazhi
[Meta-analysis on the effect and adverse reaction on patients with osteoarthritis and rheumatoid arthritis treated with non-steroidal anti-inflammatory drugs].
PMID 14687510
Abstract
To observe the rate of efficacy and adverse drug reaction of non-steroidal anti-inflammatory drugs (NSAIDs) in the population with osteoarthritis and rheumatoid arthritis, based on available clinical data. Using Meta analysis to evaluate the data of effect and safety profile of NSAIDs from 19 articles on randomized clinical trials published from 1990 to 2001 in Chinese journals. The total number of patients enrolled for evaluation on rates of effectiveness and adverse drug reaction were 1 732 and 2 925, respectively. Data on the effect and safety were comparatively heterogeneous among different kinds of NSAIDs. The effective rates (95% CI) were as follows: nabunetone, 66.7% (61.9% - 71.4%); meloxicam, 68.4% (59.2% - 77.6%); naproxen, 64.5% (59.8% - 69.1%); nimesulide, 79.8% (75.7% - 84.0%); ibuprofen, 77.2% (70.7% - 83.8%); diclofenac, 77.1% (69.2% - 85.0%); oxaprozin, 65.8% (59.5% - 72.0%). Rates of adverse drug reaction (95% CI) were as follows: nabunetone, 16.3% (12.5% - 20.0%); meloxicam, 10.2% (4.2% - 16.2%); naproxen, 29.2% (24.8% - 33.6%); nimesulide, 20.2% (16.0% - 24.3%); ibuprofen, 16.7% (14.7% - 18.8%); diclofenac, 19.3% (11.9% - 26.7%); oxaprozin, 12.7% (8.9% - 16.7%) respectively. The rates of effect and adverse reaction on patients having osteoarthritis and rheumatoid arthritis with NSAIDs treatment would largely depend on the drugs being used. Within 2 - 8 weeks of treatment, the effective rate and rate of adverse drug reaction with commonly used NSAIDs as nabumeton, meloxicam, etc., were 59.2% - 85.0% and 4.2% - 33.6%, respectively.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
O9637
Oxaprozin, solid
C18H15NO3