EMAIL THIS PAGE TO A FRIEND
Neuroscience research
Regulation of dendrite formation of Purkinje cells by serotonin through serotonin1A and serotonin2A receptors in culture.
PMID 14687886
Abstract
Serotonergic fibers and receptors appear in the rat cerebellum during early postnatal development. In the present study, we investigated the actions of serotonin (5-HT) and its receptors in the dendrite formation of Purkinje cells in organotypic cultures of anterior and posterior lobes of the cerebellum at postnatal day 7. In anterior lobes after 4 days in vitro (4DIV), the dendritic areas and branchings of Purkinje cells were increased by the treatment of 2 microM 5-HT, but decreased by 20 microM 5-HT. In posterior lobes after 4DIV, the dendritic areas of Purkinje cells were increased by 5-HT (2, 20 and 200 microM). In contrast, 5-HT treatment decreased dendritic areas of Purkinje cells in both anterior and posterior lobes after 7DIV. Next, we determined the actions of specific 5-HT receptors in mediating the effects of 5-HT by treatment with selective 5-HT receptor agonists. In anterior lobes after 4DIV, dendritic areas of Purkinje cells were increased by a 5-HT1A receptor agonist (8-OH-DPAT), whereas decreased by a 5-HT2A receptor agonist (DOI). The present study suggested that the dendrite formation of Purkinje cells is promoted by 5-HT through 5-HT1A receptors, but inhibited by 5-HT through 5-HT2A receptors.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I5763
Indophenol
C12H9NO2