EMAIL THIS PAGE TO A FRIEND
Journal of agricultural and food chemistry
Correlations between pulp properties of eucalyptus clones and leaf volatiles using automated solid-phase microextraction.
PMID 14690363
Abstract
Analysis of biogenic volatile organic compounds (BVOC) of 14 Eucalyptus clones has been performed using an automated headspace solid-phase microextraction (HS-SPME) coupled to gas chromatography (GC)/ion trap mass spectrometry (ITMS) method. Correlations between pulp properties of Eucalyptus clones and the BVOC of their leaf headspaces were studied. The compounds alpha-terpineol and the sesquiterpene beta-eudesmol were positively correlated with S5, a property related to the hemicelluose content in the pulp. Qualitative results obtained with automated HS-SPME were sufficient to group together the same species and related hybrids through cluster analysis and were confirmed through principal component analysis. A preliminary separation of the essential oils of Eucalyptus dunnii through comprehensive two-dimensional gas chromatography (GC x GC) showed approximately 580 peaks compared to approximately 60 in a typical GC/ITMS first-dimension chromatogram. The potential of HS-SPME coupled to GC x GC to improve the separation of Eucalyptus volatiles and other plant essential oils looks extremely promising for new applications of unsupervised learning methods.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
17790
β-Eudesmol, ≥90% (GC)
C15H26O