EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
3-(4-Aminobutyn-1-yl)pyridines: binding at alpha 4 beta 2 nicotinic cholinergic receptors.
PMID 14698195
Abstract
The binding of a series pyridylbutynylamines 6 was examined at alpha4beta2 nACh receptors. Structural modifications, comparing 6 with pyridyl ethers 2, did not consistently result in parallel effects on receptor affinity, suggesting possible differences in their modes of binding. Furthermore, the binding of amine 6a seemed to be accounted for by the newer vector pharmacophore models.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
130850
3-Butyn-1-ol, 97%
C4H6O