EMAIL THIS PAGE TO A FRIEND
Carbohydrate research
An efficient chemoenzymatic route to methyl 4-O-benzyl-2,3-anhydro-beta-D-lyxopyranoside from methyl beta-D-xylopyranoside.
PMID 14698903
Abstract
Methyl 4-O-benzyl-2,3-anhydro-beta-D-lyxopyranoside, an intermediate for the preparation of methyl beta-D-xylopyranoside derivatives modified at C-2, was obtained in five steps in 58% yield. The synthetic sequence starts from methyl beta-D-xylopyranoside through two main steps involving regioselective enzymatic acetylation and deacetylation catalyzed by lipase PS.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M5878
Methyl β-D-xylopyranoside, ≥99% (GC)
C6H12O5