EMAIL THIS PAGE TO A FRIEND
Proceedings of the Western Pharmacology Society
Dynamic regulation of cAMP signaling by cGMP in the cardiovascular system: roles of phosphodiesterase 2 and phosphodiesterase 3 enzymes.
PMID 14699879