EMAIL THIS PAGE TO A FRIEND
Proceedings of the Western Pharmacology Society
Opposite gender-specific increases in peripheral benzodiazepine receptor (PBR) expression in rat brain after single administration of the GABA antagonists heptachlor and heptachlor epoxide.
PMID 14699885