EMAIL THIS PAGE TO A FRIEND
Bulletin of environmental contamination and toxicology
Analysis of zinc phosphide in baits, water, soil, and biological specimens.
PMID 14705664
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
04502
Zinc phosphide, ≥19% active phoshor (P) basis, powder
P2Zn3