EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Inhibition of pancreatic adenocarcinoma cellular invasiveness by blebbistatin: a novel myosin II inhibitor.
PMID 14706640
Abstract
Blebbistatin is a novel 1-phenyl-2-pyrrolidinone derivative capable of inhibiting non-muscle myosin II activity with a high degree of specificity. We examined the effects of blebbistatin on pancreatic adenocarcinoma cellular migration, invasion, adhesion, and spreading. Blebbistatin dose-dependently inhibited cellular migration and invasiveness, quantified by modified Boyden chamber assay. Matrix metalloproteinase 2 and 9 activities were unaffected by blebbistatin and cellular proliferation was inhibited only by concentrations of blebbistatin exceeding those required to inhibit myosin II activity and to interfere with migration and invasion. While blebbistatin treatment did not affect cell adhesion to the extracellular matrix component fibronectin, it markedly impaired cell spreading on this substrate. Cell surface expression of the archetypal fibronectin receptor (alpha(5)beta(1) integrin) was unaffected by blebbistatin. Our observations illustrate the critical role of non-muscle myosin II in pancreatic adenocarcinoma cellular invasiveness and extracellular matrix interaction and suggest that therapeutic strategies targeting myosin II warrant further investigation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
307041
1-Phenyl-2-pyrrolidinone, 99%
C10H11NO