EMAIL THIS PAGE TO A FRIEND
Methods in molecular biology (Clifton, N.J.)
Analysis of mitosis in squash preparations of larval brains: orcein, Giemsa, Hoechst 33258, DAPI, quinacrine, and N-banding.
PMID 14707356