EMAIL THIS PAGE TO A FRIEND
Journal of inherited metabolic disease
3-methyglutaconic aciduria in a Chinese patient with glycogen storage disease Ib.
PMID 14707520
Abstract
We report elevated urinary excretion of 3-methylglutaconic (3MGC) and 3-methylglutaric acids (3MGR) in a patient with glycogen storage disease Ib. Combined excretion was 10-fold elevated in comparison to control during inadequate glucose maintenance, and still elevated following dietary improvement. 3MGC acid excretion correlated with plasma lactate and glucose. We speculate that imbalanced gluconeogenesis and de novo cholesterol synthesis result in secondarily increased 3MGC/3MGR production.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H4392
3-Hydroxy-3-methylglutaric acid, ≥95%
C6H10O5
M47604
3-Methylglutaric acid, 99%
C6H10O4