EMAIL THIS PAGE TO A FRIEND
Chemical & pharmaceutical bulletin
Stannyl radical addition-cyclization of oxime ethers connected with olefins.
PMID 14709871
Abstract
Stannyl radical addition-cyclization of oxime ethers connected with olefin moieties was studied. The radical reactions proceeded effectively by the use of triethylborane as a radical initiator to provide the functionalized pyrrolidines via a carbon-carbon bond-forming process.