EMAIL THIS PAGE TO A FRIEND
Methods in enzymology
Assay of intrinsic transcript termination by E. coli RNA polymerase on single-stranded and double-stranded DNA templates.
PMID 14712712
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C2258 Cellulose phosphate, (Fine Mesh)