EMAIL THIS PAGE TO A FRIEND
Advances in experimental medicine and biology
Biochemical markers predicting survival in peroxisome biogenesis disorders.
PMID 14713214
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P6617
Pristanic acid solution, mixture of isomers, ethanol solution, ≥97% (GC)
C19H38O2