EMAIL THIS PAGE TO A FRIEND
Journal of combinatorial chemistry
Stereoselective diversity-oriented solution and solid-phase synthesis of tetrahydroquinoline-based polycyclic derivatives.
PMID 14714985
Abstract
A diversity-oriented solution and solid-phase synthesis of tetrahydroquinoline-based tricyclic derivatives has been achieved from enantiomerically pure, natural product-like bicyclic scaffold. The solution synthesis of enantiopure bicyclic scaffold was developed by asymmetric hetero Michael reaction. Our approach for the synthesis of polycyclic derivatives utilized regio- and stereoselective hetero Michael reaction and ring-closing metathesis as key steps in solution and on solid phase.