EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry
Combining NMR and molecular modelling in a drug delivery context: investigation of the multi-mode inclusion of a new NPY-5 antagonist bromobenzenesulfonamide into beta-cyclodextrin.
PMID 14723963
Abstract
NMR spectroscopic and molecular modelling methods have been employed to describe the complexation of trans-N-4-[N'-(4-chlorobenzoyl)hydrazinocarbonyl]cyclohexylmethyl-4-bromobenzenesulfonamide, a new chemotype of NPY-5 antagonist, and beta-cyclodextrin, revealing the coexistence of two different kinds of 1:1 complexes where conformational changes of the guest compound with respect to the free state are also detected.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
16374
4-Bromobenzenesulfonamide, ≥99.0% (T)
C6H6BrNO2S