EMAIL THIS PAGE TO A FRIEND
Zhonghua er ke za zhi = Chinese journal of pediatrics
[Anti-inflammatory action of alpha-melanocyte-stimulating hormone].
PMID 14728905
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M4135
α-Melanocyte stimulating hormone, ≥97% (HPLC)
C77H109N21O19S