EMAIL THIS PAGE TO A FRIEND
Brain research bulletin
Chronic intracerebral infusions of vasopressin and vasopressin antagonist modulate behavioral effects of interleukin-1 in rat.
PMID 1473021
Abstract
To assess the role of sex dependent brain vasopressinergic transmission in the modulation of the neural effects of interleukin-1, castrated male rats that are deficient in vasopressin were implanted intracerebroventricularly with an Accurel collodion mini device containing 10 micrograms AVP whereas intact male rats were implanted with a similar device containing 50 micrograms of dPTyr(Me)AVP, a specific antagonist of the vasopressor-like receptors of vasopressin. Control rats in each sex group were implanted with an Accurel device containing distilled water. Acute intracerebroventricular injection of 1.25-2.50 ng recombinant human interleukin-1 beta decreased in a dose and time-dependent manner social investigation of a juvenile conspecific. This effect was more intense in intact rats chronically infused with dPTyr(Me)AVP and less intense in castrates infused with AVP. These results confirm the modulatory role of sex-dependent vasopressinergic neurotransmission on the neural effects of interleukin-1.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
V1880
[deamino-Pen1, O-Me-Tyr2, Arg8]-Vasopressin, ≥97% (HPLC)
C49H70N14O12S2