EMAIL THIS PAGE TO A FRIEND
IDrugs : the investigational drugs journal
BNP-7787 (BioNumerik/Baxter Oncology/Grelan).
PMID 14730469
Abstract
BioNumerik Pharmacology Inc, Baxter Oncology GmbH (formerly ASTA Medica AG) and Grelan Pharmaceutical Co Ltd are developing BNP-7787 for the potential reduction of toxicity associated with cisplatin and carboplatin treatment in cancer patients.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Y0000317 Mesna impurity D, European Pharmacopoeia (EP) Reference Standard
C4H10O6S4