EMAIL THIS PAGE TO A FRIEND
Journal of agricultural and food chemistry
Liquid chromatographic determination of fumonisins B1, B2, and B3 in corn silage.
PMID 14733494
Abstract
Corn silage was dried, ground, and then extracted with 0.1 M ethylenediaminetetraacetic acid. The filtrate was applied to a FumoniTest immunoaffinity column. Fumonisins were derivatized with naphthalene-2,3-dicarboxaldehyde, separated on a C(18) liquid chromatographic column, and detected by fluorescence. The detection limits for fumonisin B(1), fumonisin B(2), and fumonisin B(3) were 50, 25, and 25 ng/g of dried silage, respectively. Recoveries of fumonisin B(1), fumonisin B(2), and fumonisin B(3) from wet and dried corn silage spiked over the range of 100-5000 ng/g averaged 91-106%. The method was applied to corn silage samples collected from the midwestern area of the United States during 2001-2002. Of 89 corn silage samples, fumonisin B(1), fumonisin B(2), and fumonisin B(3) were found in 86 (97%), 64 (72%), and 51 (57%) of the samples. The mean positive levels of fumonisin B(1), fumonisin B(2), and fumonisin B(3) were 615, 93, and 51 ng/g, respectively, in dried silage. This suggests that fumonisins may be frequent low level contaminants in corn silage.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
32606
Fumonisin B3 solution, 50 μg/mL in acetonitrile: water, analytical standard
C34H59NO14