EMAIL THIS PAGE TO A FRIEND
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association
Toxicology and hazard potential of trifluralin.
PMID 1473797
Abstract
This paper reviews the results of toxicity studies conducted in laboratory animals to evaluate the safety of the herbicide trifluralin (TFL). The data show that TFL is slightly toxic following single oral exposure. Testing for embryotoxicity in rats and rabbits indicated no teratogenic potential, and many different mutagenicity tests showed that TFL was non-genotoxic. Subchronic and chronic toxicity testing in rats, mice and dogs indicated that TFL was haematotoxic (anaemia and methaemoglobinaemia), particularly in the dog, and slightly hepatotoxic. No-observed-effect levels of 4.8 and 41 mg/kg body weight/day, respectively, were determined in dogs and rats exposed chronically to TFL. Oncogenicity studies in rats and mice revealed no carcinogenic potential. Since the data for TFL indicated no mutagenic or other special toxicological risks, it is suggested that a safety factor of 100 could be used for the determination of the acceptable daily intake of TFL, which would be 0.05 mg/kg body weight/day.