EMAIL THIS PAGE TO A FRIEND
Der Hautarzt; Zeitschrift fur Dermatologie, Venerologie, und verwandte Gebiete
[Topical use of inorganic mercury compounds].
PMID 1473976
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
10005
Mercury(II) amidochloride, ≥95.0%, powder
H2ClHgN