EMAIL THIS PAGE TO A FRIEND
The Journal of antibiotics
Asterric acid, a new endothelin binding inhibitor.
PMID 1473998
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SMB00012
Asterric acid, ≥95% (LC/MS-ELSD)
C17H16O8