EMAIL THIS PAGE TO A FRIEND
European journal of clinical pharmacology
Phenotyping of N-acetyltransferase type 2 by caffeine from uncontrolled dietary exposure.
PMID 14747882
Abstract
The standard approach for phenotyping of the human arylamine N-acetyltransferase 2 (NAT2) uses urinary caffeine metabolite ratios after a caffeine test dose taken in after methylxanthine abstinence. We tested whether these standardization measures were still needed when a more sensitive quantification technique was used. A new liquid chromatography/tandem mass spectrometry (LC-MS/MS) method for the quantification of the caffeine metabolites 5-acetylamino-6-formylamino-3-methyluracil (AFMU), 5-acetylamino-6-amino-3-methyluracil (AAMU), 1-methylxanthine (1X), and 1-methylurate (1U) was developed. Urine samples from 77 healthy volunteers collected before and 5-6 h after oral intake of 150-200 mg caffeine were analyzed. The lower limits of quantification were 0.1 microg/ml for caffeine, 1X, 1U, and AFMU, and 0.2 microg/ml for AAMU. The urinary NAT2 ratios (AFMU+AAMU) / (AFMU+AAMU+1X+1U) before and after caffeine intake correlated well in 65 volunteers (r(2)=0.827; P< 0.0001). In 12 participants (16%), metabolite concentrations in urine before caffeine intake were below the quantification limit. NAT2 genotyping, done in 41 volunteers for four SNPs, corroborated the phenotyping results. NAT2 activity can be determined from a spontaneous urine probe in most subjects by quantification of caffeine metabolites arising from non-standardized dietary caffeine exposure using LC-MS/MS. This may facilitate the phenotyping procedure.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M6885
1-Methyluric acid
C6H6N4O3
UC432
AAMU
C7H10N4O3