EMAIL THIS PAGE TO A FRIEND
Mycoses
The effect of sulconazole on the antigenic composition of two dermatophyte species.
PMID 1474985
Abstract
Trichophyton rubrum and Trichophyton interdigitale have been grown in liquid culture in the presence of sulconazole. The antigenic activity of detergent extracts of intact organisms was analysed following SDS-PAGE and the probing of Western blots with homologous antisera raised in rabbits and with sera from patients with dermatophyte infections. Differences in protein-band patterns were noted; some bands present in control samples were absent in azole-treated samples and vice versa. These differences were reflected in antigenic band patterns, especially among components of approximate molecular weight of 30-40, 50-60 and 92-100 kDa.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S9632
Sulconazole nitrate salt
C18H15Cl3N2S · HNO3