EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences
Urine sample preparation of tricyclic antidepressants by means of a supported liquid membrane technique for high-performance liquid chromatographic analysis.
PMID 14751789
Abstract
Supported liquid membrane (SLM) technique for sample work-up and enrichment was used for determination of tricyclic antidepressant drugs in urine by high-performance liquid chromatography (HPLC) with UV detection. The studied antidepressant drugs were amitriptyline, opipramol, noxiptyline and additionally diethazine was used as possible internal standard. Alkaline phosphoric buffer with urine sample, as the donor solution, was passed over the liquid membrane into which investigated substances were extracted. On the other side of the membrane, analyzed compounds were trapped due to creating non-extractable form in acidic acceptor solution. Enriched and cleaned up drugs were then injected into a HPLC system with ultraviolet detection to analyze of their concentration in acceptor solution. Optimum extraction efficiency was determined by changing acceptor and donor solutions pH, application of different flow rates of donor solution and by using different solvents in the membrane. Also, donor solution volume, extraction time and concentration of analytes were varied to check the linearity of extraction process. The highest extraction efficiency: 43% for opipramol, 56% for noxiptyline, 43% for amitriptyline and 42% for diethazine (R.S.D. values were <6% and n=3) was achieved when 0.05 M phosphate buffer pH 4.0 and 9.5 were used as donor and acceptor solutions, respectively, n-undecane with 5% tri-n-octylphosphine oxide (TOPO) was used as liquid membrane. Limit of quantification (LOQ) for tricyclic antidepressants after enrichment of 100ml of urine sample was about 1 ng/ml.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
O5889
Opipramol dihydrochloride, ≥98% (HPLC), solid
C23H29N3O · 2HCl