EMAIL THIS PAGE TO A FRIEND
Journal of Translational Medicine
A novel technique for quantifying changes in vascular density, endothelial cell proliferation and protein expression in response to modulators of angiogenesis using the chick chorioallantoic membrane (CAM) assay.
PMID 14754458