EMAIL THIS PAGE TO A FRIEND
Proceedings of the National Academy of Sciences of the USA
The gene for soluble N-ethylmaleimide sensitive factor attachment protein alpha is mutated in hydrocephaly with hop gait (hyh) mice.
PMID 14755058