EMAIL THIS PAGE TO A FRIEND
Regulatory peptides
The cholecystokinin2-receptor mediates calcitonin secretion, gene expression, and proliferation in the human medullary thyroid carcinoma cell line, TT.
PMID 14759564
Abstract
Gastrin-induced release of calcitonin from medullary thyroid carcinomas (MTC) is based on the expression of the cholecystokinin(2)-receptor (CCK(2)R) in these tumors. Recently, we have shown that the CCK(2)R is expressed not only in MTC but also in C-cells within the normal thyroid gland. The functions of the CCK(2)R in MTC and C-cells are largely unknown. We therefore explored the effects of gastrin-induced CCK(2)R stimulation in the highly differentiated MTC cell line, TT. CCK(2)R expression in TT-cells is detectable by RT-PCR as well as immunocytochemistry. Stimulation of the CCK(2)R by gastrin induces immediate release of calcitonin from TT-cells. Moreover, quantitative (LightCycler) RT-PCR demonstrates that gastrin stimulates transcription of the calcitonin and chromogranin A genes in TT-cells. TT-cell proliferation, assessed by counting of viable cells and (3)H-thymidine uptake, is markedly increased by gastrin. This effect is inhibited by the CCK(2)R-specific antagonist L-365,260. Our findings suggest physiological functions for the CCK(2)R in calcitonin-secretion and gene expression as well as a pathophysiological role in MTC proliferation. CCK(2)R antagonists might have therapeutic potential in these tumors.